Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethyl malonate ≥98.0% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: Malonic acid diethyl ester1,3-Propanedioic acid diethyl ester

M-Clarity™ quality level = MQ200

8.00898.1000 8.00898.0100 8.00898.0500 8.00898.2500 8.00898.0100 8.00898.2500 8.00898.1000 8.00898.0500
EM8.00898.1000CS 553 USD InStock
CA8.00898.0500 EM8.00898.1000 EM8.00898.0100 CA8.00898.1000 EM8.00898.2500 EM8.00898.0500 CA8.00898.2500 CA8.00898.0100
Diethyl malonate ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
Diethyl malonate


Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 199 °C (1013 hPa)
Melting Pt: –50 °C
Density: 1.055 g/cm³ (20 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00009195
CAS Number: 105-53-3
EINECS: 203-305-9
Merck Index: 13,03856
Order Now
Specification Test Results
Assay (GC, area%) ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C) 1.053 - 1.057
Identity (IR) passes test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty