Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethylenediamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: 1,2-Diaminoethane1,2-Ethanediamine

M-Clarity™ quality level = MQ200

8.00947.0500 8.00947.1000 8.00947.0100 8.00947.2500 8.00947.0100 8.00947.2500 8.00947.1000 8.00947.9051 8.00947.0500
EM8.00947.1000CS 581 USD InStock
EM8.00947.1000 CA8.00947.0500 CA8.00947.0100 CA8.00947.2500 EM8.00947.0500 CA8.00947.1000 CA8.00947.9051 EM8.00947.2500 EM8.00947.0100
Ethylenediamine for synthesis, Sigma-Aldrich®
Ethylenediamine


Formula: C₂H₈N₂
MW: 60.1 g/mol
Boiling Pt: 117 °C (1013 hPa)
Melting Pt: 8.5 °C
Density: 0.898 g/cm³ (20 °C)
Flash Pt: 34 °C
Storage Temperature: Ambient
MDL Number: MFCD00008204
CAS Number: 107-15-3
EINECS: 203-468-6
UN: 1604
ADR: 8,II
Merck Index: 13,03829
https://us-legacy.vwr.com/stibo/web/std.lang.all/99/69/2299969.jpg
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point 11 °C
Molar Mass 60.10 g/mol
Boiling Point 116 °C (1013 hPa)
Vapor Pressure 12 hPa (20 °C)
Flash Point 38 °C
Explosion Limit 2.5 - 16.3 %(V)
Density 0.897 g/cm3 (20 °C)
pH 12.2 (100 g/l, H2O, 20 °C)
Ignition Point 405 °C
Hazard Class 8
Hazard Class Description Corrosive material

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty