Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium isopropoxide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: AluminiumtriisopropylateAluminium isopropoxideAluminiumtriisopropoxideAluminiumisopropylate

M-Clarity™ quality level = MQ200

8.01079.1000 8.01079.1000 8.01079.0250 8.01079.0250
EM8.01079.0250CS 740 USD InStock
CA8.01079.1000 CA8.01079.0250 EM8.01079.0250 EM8.01079.1000
Aluminium isopropoxide for synthesis, Sigma-Aldrich®
Aluminium isopropoxide


Formula: C₉H₂₁AlO₃
MW: 204.25 g/mol
Boiling Pt: 135 °C (13 hPa)
Melting Pt: 119 °C
Density: 0.9771 g/cm³ (20 °C)
MDL Number: MFCD00008870
CAS Number: 555-31-7
EINECS: 209-090-8
UN: 3181
ADR: 4.1,II
Merck Index: 13,00346
Order Now
Specification Test Results
Solubility (20 °C) (decomposition)
Melting Point 130 - 134 °C
Molar Mass 204.24 g/mol
Boiling Point 94 °C (0.6 hPa)
Vapor Pressure 0.13 hPa (21 °C)
Flash Point 26 °C
Refractive Index 1.432 (23 °C, 589 nm)
Explosion Limit 2 %(V)
Density 1.035 g/cm3 (20 °C)
Hazard Class 4.1
Hazard Class Description Flammable Solid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty