Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

9-Aminoacridine hydrochloride monohydrate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: 9-Aminoacridine hydrochloride hydrateAcridin-9-amine hydrochloride hydrate

M-Clarity™ quality level = MQ200

8.01112.0100 8.01112.0100 8.01112.0025 8.01112.0002 8.01112.0025
EM8.01112.0025CS 11554 USD InStock
EM8.01112.0025 EM8.01112.0002 EM8.01112.0100 CA8.01112.0025 CA8.01112.0100
9-Aminoacridine hydrochloride monohydrate for synthesis, Sigma-Aldrich®
9-Aminoacridine hydrochloride monohydrate


Formula: C₁₃H₁₁ClN₂·1H₂O
MW: 248.71 g/mol
MDL Number: MFCD00150071
CAS Number: 52417-22-8
EINECS: 205-145-5
ADR: 6.1,III
Order Now
Specification Test Results
Solubility 3.3 g/l
Melting Point 240 - 244 °C
Molar Mass 248.71 g/mol

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty