Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethylbenzene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: Phenylethane

M-Clarity™ quality level = MQ200

8.01372.9045 8.01372.0100 8.01372.9045 8.01372.0100 8.01372.1000 8.01372.1000
EM8.01372.0100CS 728.97 USD InStock
CA8.01372.0100 CA8.01372.1000 EM8.01372.0100 CA8.01372.9045 EM8.01372.1000 EM8.01372.9045
Ethylbenzene for synthesis, Sigma-Aldrich®
Ethylbenzene


Formula: C₈H₁₀
MW: 106.17 g/mol
Boiling Pt: 136.2 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00011647
CAS Number: 100-41-4
EINECS: 202-849-4
UN: 1175
ADR: 3,II
Merck Index: 13,03801
https://us-legacy.vwr.com/stibo/web/std.lang.all/60/41/766041.jpg
Order Now
Specification Test Results
Solubility 0.2 g/l (20 °C)
Melting Point -95 °C
Molar Mass 106.16 g/mol
Boiling Point 136 °C (1013 hPa)
Vapor Pressure 9.5 hPa (20 °C)
Flash Point 15 °C
Explosion Limit 1.0 - 7.8 %(V)
Density 0.87 g/cm3 (20 °C)
Ignition Point 430 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty