Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aminoguanidinium hydrogen carbonate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: 1-Aminoguanidinium hydrogen carbonateAminoguanidine hydrogen carbonate

M-Clarity™ quality level = MQ200

8.01417.0250 8.01417.0250
EM8.01417.0250CS 1472 USD InStock
EM8.01417.0250 CA8.01417.0250
Aminoguanidinium hydrogen carbonate for synthesis, Sigma-Aldrich®
Aminoguanidinium hydrogen carbonate


Formula: C₂H₈N₄O₃
MW: 136.11 g/mol
MDL Number: MFCD00012949
CAS Number: 2582-30-1
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Solubility  - 5 g/l (20 °C)
Melting Point 125 °C decomposes
Molar Mass 136.11 g/mol
Bulk Density 700 kg/m3
Density 1.56 g/cm3 (20 °C)
pH 8.9 (5 g/l, H2O, 20 °C)
Ignition Point 245 °C
Synonyms Aminoguanidine bicarbonate
Hill Formula C2H8N4O3
Chemical Formula CH6N4 * H2CO3

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty