Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

AMP (2-Amino-2-methylpropanol) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: 2-Amino-2-methyl-1-propanolAMP

M-Clarity™ quality level = MQ200

8.01465.0250 8.01465.1000 8.01465.9026 8.01465.1000 8.01465.2500 8.01465.0250 8.01465.9026 8.01465.2500
EM8.01465.9026EA 1220 USD InStock
EM8.01465.9026 CA8.01465.2500 EM8.01465.0250 EM8.01465.1000 EM8.01465.2500 CA8.01465.0250 CA8.01465.1000 CA8.01465.9026
AMP (2-Amino-2-methylpropanol) for synthesis, Sigma-Aldrich®
AMP (2-Amino-2-methylpropanol)


Formula: (CH₃)₂C(NH₂)CH₂OH
MW: 89.14 g/mol
Boiling Pt: 165 °C (1013 hPa)
Melting Pt: 18…26 °C
Density: 0.667 g/cm³ (20 °C)
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00008051
CAS Number: 124-68-5
EINECS: 204-709-8
Merck Index: 13,00448
Order Now
Specification Test Results
Melting Point 30 - 31 °C
Molar Mass 89.14 g/mol
Boiling Point 165 °C (1013 hPa) (anhydrous)
Vapor Pressure 0.45 hPa (20 °C)
Flash Point 77.8 °C
Refractive Index 1.448 (20 °C, 589 nm)
Density 0.95 g/cm3 (25 °C)
pH 11.3 (10 g/l, H2O)
Ignition Point 438 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty