Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Butylamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: n-Butylamine1-Aminobutane1-Butylamine

M-Clarity™ quality level = MQ200

8.01539.2500 8.01539.1000 8.01539.1000 8.01539.2500
EM8.01539.1000CS 405 USD InStock
EM8.01539.1000 CA8.01539.1000 EM8.01539.2500 CA8.01539.2500
Butylamine for synthesis, Sigma-Aldrich®
Butylamine


Formula: H₃C(CH₂)₂CH₂NH₂
MW: 73.14 g/mol
Boiling Pt: 77 °C (1013 hPa)
Melting Pt: –50 °C
Density: 0.741 g/cm³ (20 °C)
Flash Pt: –8 °C
Storage Temperature: Ambient
MDL Number: MFCD00011690
CAS Number: 109-73-9
EINECS: 203-699-2
UN: 1125
ADR: 3 (8),II
Merck Index: 12,01578
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point -50 °C
Molar Mass 73.14 g/mol
Boiling Point 76 - 78 °C
Vapor Pressure 93 hPa (20 °C)
Flash Point -8.5 °C DIN 51755 Part 1
Refractive Index 1.4015 (20 °C, 589 nm)
Explosion Limit 1.5 - 9.8 %(V)
Density 0.74 g/cm3 (20 °C)
pH 12.6 (100 g/l, H2O, 20 °C)
Ignition Point 290 °C
Hazard Class 3
Hazard Class Description Flammable liquid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty