Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzophenone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: DiphenylmethanoneDiphenyl ketone

M-Clarity™ quality level = MQ200

8.01801.0005 8.01801.0005
EM8.01801.0005CS 15525 USD InStock
CA8.01801.0005 EM8.01801.0005
Benzophenone for synthesis, Sigma-Aldrich®
Benzophenone


Formula: C₁₃H₁₀O
MW: 182.22 g/mol
Boiling Pt: 305.4 °C (1013 hPa)
Melting Pt: 47…49 °C
Density: 1.085 g/cm³ (20 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00003076
CAS Number: 119-61-9
EINECS: 204-337-6
UN: 3077
ADR: 9,III
Merck Index: 13,01099
Order Now
Specification Test Results
Solubility 0.14 g/l (25 °C)
Melting Point 47 - 49 °C
Molar Mass 182.22 g/mol
Bulk Density 700 kg/m3
Boiling Point 304 - 306 °C (1013 hPa)
Vapor Pressure 0.004 hPa (20 °C)
Flash Point 150 °C
Density 1.1 g/cm3 (25 °C)
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty