Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzyl chloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: (Chloromethyl)benzeneChlorophenylmethaneα-Chlorotoluene

M-Clarity™ quality level = MQ200

8.01809.0100 8.01809.2500 8.01809.1000 8.01809.2500 8.01809.1000 8.01809.0100
EM8.01809.1000CS 306 USD InStock
CA8.01809.0100 CA8.01809.2500 CA8.01809.1000 EM8.01809.1000 EM8.01809.2500 EM8.01809.0100
Benzyl chloride for synthesis, Sigma-Aldrich®
Benzyl chloride


Formula: (C₆H₅)CH₂Cl
MW: 126.59 g/mol
Boiling Pt: 179.3 °C (1013 hPa)
Melting Pt: –41.2 °C
Density: 1.103 g/cm³ (20 °C)
Flash Pt: 52 °C
Storage Temperature: Ambient
MDL Number: MFCD00000889
CAS Number: 100-44-7
EINECS: 202-853-6
UN: 1738
ADR: 6.1 (8),II
Merck Index: 13,01131
Order Now
Specification Test Results
Solubility 460 mg/l (30 °C)
Melting Point -41.2 °C
Molar Mass 126.58 g/mol
Boiling Point 179.3 °C (1013 hPa)
Vapor Pressure 1.2 hPa (20 °C)
Flash Point 67 °C
Refractive Index 1.5415
Explosion Limit 1.1 - 14 %(V)
Density 1.10 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 585 °C
Hazard Class 6.1
Hazard Class Description Toxic materials

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty