Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Ethoxyphenol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: Pyrocatechol monoethyl ethero-Ethoxyphenol1-Hydroxy-2-ethoxybenzene

M-Clarity™ quality level = MQ200

8.01917.0250 8.01917.0250
EM8.01917.0250CS 685 USD InStock
CA8.01917.0250 EM8.01917.0250
2-Ethoxyphenol for synthesis, Sigma-Aldrich®
2-Ethoxyphenol


Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 214…216 °C (1013 hPa)
Melting Pt: 28…29 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00002187
CAS Number: 94-71-3
EINECS: 202-358-5
Order Now
Specification Test Results
Solubility 9 g/l (30 °C)
Melting Point 24 - 28 °C
Molar Mass 138.16 g/mol
Boiling Point 214 - 216 °C (1013 hPa)
Vapor Pressure 1 hPa (45 °C)
Flash Point 86 °C
Refractive Index 1.5291 (25 °C, 589 nm)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty