Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chlorophenol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: 1-Chloro-2-hydroxybenzene

M-Clarity™ quality level = MQ200

8.02253.0500 8.02253.2500 8.02253.0100 8.02253.0100 8.02253.0500 8.02253.2500
EM8.02253.0500CS 372 USD InStock
EM8.02253.0500 EM8.02253.2500 CA8.02253.2500 CA8.02253.0100 CA8.02253.0500 EM8.02253.0100
2-Chlorophenol for synthesis, Sigma-Aldrich®
2-Chlorophenol


Formula: C₆H₅ClO
MW: 128.56 g/mol
Boiling Pt: 175 °C (1013 hPa)
Melting Pt: 9 °C
Density: 1.25 g/cm³ (20 °C)
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00002159
CAS Number: 95-57-8
EINECS: 202-433-2
UN: 2021
ADR: 6.1,III
Merck Index: 12,02173
Order Now
Specification Test Results
Solubility 28.5 g/l (20 °C)
Melting Point 7 °C
Molar Mass 128.55 g/mol
Boiling Point 174 °C (1013 hPa)
Vapor Pressure 13.3 hPa (50 °C)
Flash Point 63.9 °C
Refractive Index 1.5579 (20 °C, 589 nm)
Density 1.26 g/cm3 (20 °C)
Ignition Point 550 °C
Hazard Class 6.1
Hazard Class Description Toxic Materials

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty