Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloroaniline for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

M-Clarity™ quality level = MQ200

8.02611.0100 8.02611.0100 8.02611.0500 8.02611.0500
EM8.02611.0500CS 1044 USD InStock
CA8.02611.0500 EM8.02611.0500 CA8.02611.0100 EM8.02611.0100
2-Chloroaniline for synthesis, Sigma-Aldrich®
2-Chloroaniline


Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 208 °C (1013 hPa)
Melting Pt: –3 °C
Density: 1.213 g/cm³ (20 °C)
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00007656
CAS Number: 95-51-2
EINECS: 202-426-4
UN: 2019
ADR: 6.1,II
Merck Index: 13,02136
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point -3 °C
Molar Mass 127.57 g/mol
Boiling Point 207 - 209 °C (1013 hPa)
Vapor Pressure 0.5 hPa (20 °C)
Flash Point 105 °C
Refractive Index 1.5877 (20 °C, 589 nm)
Explosion Limit 2.4 - 14.2 %(V)
Density 1.21 g/cm3 (20 °C)
pH (H2O, 20 °C) suspension,weakly alkaline
Ignition Point 500 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty