Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Crotonic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: 2-Butenoic acidtrans-2-Butenoic acidtrans-Crotonic acid

M-Clarity™ quality level = MQ200

8.02650.1000 8.02650.0100 8.02650.1000 8.02650.0100
EM8.02650.1000CS 847 USD InStock
EM8.02650.1000 EM8.02650.0100 CA8.02650.1000 CA8.02650.0100
Crotonic acid for synthesis, Sigma-Aldrich®
Crotonic acid


Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 185…199 °C (1013 hPa)
Melting Pt: 71…72 °C
Density: 1.02 g/cm³ (20 °C)
Flash Pt: 90 °C
MDL Number: MFCD00002701
CAS Number: 107-93-7
EINECS: 203-533-9
UN: 2823
ADR: 8,III
Merck Index: 13,02625
Order Now
Specification Test Results
Solubility 6.2 g/l (20 °C)
Melting Point 70 - 73 °C
Molar Mass 86.09 g/mol
Bulk Density 543 kg/m3
Boiling Point 185.0 - 199.5 °C (1013 hPa)
Vapor Pressure 0.25 hPa (20 °C)
Flash Point 90 °C
Density 1.02 g/cm3 (20 °C)
pH 3 (10 g/l, H2O, 20 °C)
Ignition Point 500 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty