Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclopentanone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

M-Clarity™ quality level = MQ200

8.02670.1000 8.02670.9025 8.02670.0250 8.02670.1000 8.02670.2500 8.02670.0250 8.02670.2500
EM8.02670.9025EA 1679 USD InStock
EM8.02670.9025 CA8.02670.1000 EM8.02670.2500 CA8.02670.0250 EM8.02670.0250 EM8.02670.1000 CA8.02670.2500
Cyclopentanone for synthesis, Sigma-Aldrich®
Cyclopentanone


Formula: C₅H₈O
MW: 84.12 g/mol
MDL Number: MFCD00001409
CAS Number: 120-92-3
UN: 2245
ADR: 3,III
Order Now
Specification Test Results
Solubility (20 °C) slightly soluble
Melting Point -58 °C
Molar Mass 84.11 g/mol
Boiling Point 131 °C (1013 hPa)
Vapor Pressure 11.5 hPa (20 °C)
Flash Point 27 °C
Explosion Limit 1.6 - 10.8 %(V)
Density 0.95 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 445 °C
Hill Formula C5H8O

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty