Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bis(2-ethoxyethyl)ether for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: DiethyleneglycoldiethyletherDiethylene glycol diethyl ether2-Ethoxyethyl ether

M-Clarity™ quality level = MQ200

8.02932.1000 8.02932.1000 8.02932.0250 8.02932.0250
EM8.02932.0250CS 673 USD InStock
CA8.02932.0250 CA8.02932.1000 EM8.02932.0250 EM8.02932.1000
Bis(2-ethoxyethyl)ether for synthesis, Sigma-Aldrich®
Bis(2-ethoxyethyl) ether


Formula: C₈H₁₈O₃
MW: 162.23 g/mol
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: –44 °C
Density: 0.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009254
CAS Number: 112-36-7
EINECS: 203-963-7
Merck Index: 13,03145
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point -44 °C
Molar Mass 162.22 g/mol
Boiling Point 189 °C (1013 hPa)
Vapor Pressure 0.5 hPa (20 °C)
Flash Point 71 °C
Refractive Index 1.4115 (20 °C, 589 nm)
Density 0.91 g/cm3 (20 °C)
Ignition Point 174 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty