Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3-Dibromo-5,5-dimethylhydantoin for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: 1,3-Dibromo-5,5-dimethylimidazolidine-2,4-dione

M-Clarity™ quality level = MQ200

8.03600.0250 8.03600.0250
EM8.03600.0250CS 937 USD InStock
CA8.03600.0250 EM8.03600.0250
1,3-Dibromo-5,5-dimethylhydantoin for synthesis, Sigma-Aldrich®
1,3-Dibromo-5,5-dimethylhydantoin


Formula: C₅H₆Br₂N₂O₂
MW: 285.92 g/mol
MDL Number: MFCD00003189
CAS Number: 77-48-5
EINECS: 201-030-9
UN: 3084
ADR: 8 (5.1),II
Order Now
Specification Test Results
Solubility 1 g/l (20 °C)
Melting Point 186.0 - 192.0 °C
Molar Mass 285.92 g/mol
Flash Point 155.0 °C
Density 1.00 g/cm3 (25 °C)
Ignition Point 550 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty