Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glycolic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: 2-Hydroxy acid2-Hydroxyacetic acid

M-Clarity™ quality level = MQ200

8.04104.0100 8.04104.0100 8.04104.0250 8.04104.1000 8.04104.1000 8.04104.0250
EM8.04104.0100CS 19247 USD InStock
CA8.04104.1000 CA8.04104.0250 EM8.04104.0100 EM8.04104.0250 EM8.04104.1000 CA8.04104.0100
Glycolic acid for synthesis, Sigma-Aldrich®
Glycolic acid


Formula: C₂H₄O₃
MW: 76.05 g/mol
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: 79…80 °C
Density: 1.49 g/cm³ (20 °C)
Flash Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00004312
CAS Number: 79-14-1
EINECS: 201-180-5
UN: 3261
ADR: 8,II
Merck Index: 13,04511
Order Now
Specification Test Results
Assay (acidimetric) ≥ 99.0 %
Identity (IR) passes test
Melting range (lower value) ≥ 74 °C
Melting range (upper value) ≤ 79 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty