Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Hydroxybenzaldehyde for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

M-Clarity™ quality level = MQ200

8.04536.0250 8.04536.0250 8.04536.5000 8.04536.0100 8.04536.0100 8.04536.5000
EM8.04536.5000EA 839 USD InStock
EM8.04536.5000 EM8.04536.0250 EM8.04536.0100 CA8.04536.0250 CA8.04536.5000 CA8.04536.0100
4-Hydroxybenzaldehyde for synthesis, Sigma-Aldrich®
4-Hydroxybenzaldehyde


Formula: C₇H₆O₂
MW: 122.12 g/mol
MDL Number: MFCD00006939
CAS Number: 123-08-0
Order Now
Specification Test Results
Solubility 13.8 g/l (30.5 °C)
Melting Point 116 °C
Molar Mass 122.12 g/mol
Boiling Point (1013 hPa) (sublimed)
Flash Point 174 °C
Density 1.13 g/cm3 (20 °C)
Ignition Point 450 °C
Hill Formula C7H6O2
Chemical Formula 4-(OH)C6H4CHO

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty