Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrazinium hydroxide ∼ 100% N₂H₅OH for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: Hydrazine hydrateHydrazine monohydrate

M-Clarity™ quality level = MQ200

8.04608.9055 8.04608.1000 8.04608.0250 8.04608.1000 8.04608.0250 8.04608.9055
EM8.04608.9055EA 2139 USD InStock
CA8.04608.9055 EM8.04608.9055 EM8.04608.0250 EM8.04608.1000 CA8.04608.0250 CA8.04608.1000
Hydrazinium hydroxide ∼ 100% N₂H₅OH for synthesis, Sigma-Aldrich®
Hydrazinium hydroxide


Formula: NH₂NH₂·H₂O
MW: 50.06 g/mol
Boiling Pt: 117.2 °C (1013 hPa)
Melting Pt: –51.7 °C
Density: 1.03 g/cm³ (20 °C)
Flash Pt: 75 °C
MDL Number: MFCD00149931
CAS Number: 7803-57-8
EINECS: 206-114-9
UN: 2030
ADR: 8,II
Merck Index: 13,04790
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point -51.7 °C
Molar Mass 50.06 g/mol
Boiling Point 120.5 °C (1013 hPa)
Vapor Pressure 20 hPa (20 °C)
Flash Point 33 °C
Explosion Limit 3.4 - 99 %(V)
Density 1.03 g/cm3 (20 °C)
pH 12 (640 g/l, H2O, 20 °C)
Ignition Point 280 °C
Hazard Class 8
Hazard Class Description Corrosive Material

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty