Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-2-Methyl-1-butanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

M-Clarity™ quality level = MQ200

8.06031.1000 8.06031.0100 8.06031.0500 8.06031.1000 8.06031.0500 8.06031.0100
EM8.06031.0500CS 1183 USD InStock
CA8.06031.0100 EM8.06031.0500 CA8.06031.0500 CA8.06031.1000 EM8.06031.0100 EM8.06031.1000
(±)-2-Methyl-1-butanol for synthesis, Sigma-Aldrich®
(±)-2-Methyl-1-butanol


Formula: C₅H₁₂O
MW: 88.15 g/mol
MDL Number: MFCD00004743
CAS Number: 137-32-6
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Solubility 36 g/l (20 °C)
Melting Point -70 °C
Molar Mass 88.15 g/mol
Boiling Point 127 - 129 °C
Vapor Pressure 4.3 hPa (20 °C)
Flash Point 42.5 °C
Refractive Index 1.4107 (20 °C, 589 nm)
Explosion Limit 1.2 - 10.3 %(V)
Density 0.82 g/cm3 (20 °C)
pH 7 (H2O)
Ignition Point 340 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty