Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Morpholine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger

M-Clarity™ quality level = MQ200

8.06127.1000 8.06127.2500 8.06127.2500 8.06127.0100 8.06127.0100 8.06127.0500 8.06127.9025 8.06127.1000 8.06127.0500 8.06127.9025
EM8.06127.9025EA 675 USD InStock
CA8.06127.0100 CA8.06127.2500 EM8.06127.9025 EM8.06127.0500 CA8.06127.0500 CA8.06127.1000 EM8.06127.2500 CA8.06127.9025 EM8.06127.0100 EM8.06127.1000
Morpholine for synthesis, Sigma-Aldrich®
Morpholine


Formula: C₄H₉NO
MW: 87.12 g/mol
Boiling Pt: 128.9 °C (1013 hPa)
Melting Pt: –5 °C
Density: 1 g/cm³ (20 °C)
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00005972
CAS Number: 110-91-8
EINECS: 203-815-1
UN: 2054
ADR: 8 (3),I
Merck Index: 12,06362
https://us-legacy.vwr.com/stibo/web/std.lang.all/82/95/2298295.jpg
Order Now
Specification Test Results
Solubility (20 °C) soluble
Molar Mass 87.12 g/mol
Boiling Point 128 - 130 °C (1013 hPa)
Vapor Pressure 10 hPa (20 °C)
Flash Point 32 °C
Density 1.00 g/cm3 (20 °C)
pH 10.6 (5 g/l, H2O, 20 °C)
Ignition Point 275 °C
Hazard Class 8
Hazard Class Description Corrosive Material

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty