Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromonaphthalene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: α-Bromonaphthalene

M-Clarity™ quality level = MQ200

8.06210.0100 8.06210.2500 8.06210.0100 8.06210.0500 8.06210.0500 8.06210.2500
EM8.06210.2500CS 6188 USD InStock
CA8.06210.0500 CA8.06210.0100 CA8.06210.2500 EM8.06210.2500 EM8.06210.0500 EM8.06210.0100
1-Bromonaphthalene for synthesis, Sigma-Aldrich®
1-Bromonaphthalene


Formula: C₁₀H₇Br
MW: 207.07 g/mol
MDL Number: MFCD00003868
CAS Number: 90-11-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/85/39/788539.jpg
Order Now
Specification Test Results
Solubility (20 °C) practically insoluble
Molar Mass 207.07 g/mol
Boiling Point 281 °C (1013 hPa)
Vapor Pressure 0.013 hPa (20 °C)
Flash Point 67 °C
Refractive Index 1.6576 (20 °C, 589 nm)
Density 1.48 g/cm3 (20 °C)
Synonyms α-Naphthyl bromide
Hill Formula C10H7Br

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty