Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphorus, red stabilized for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

M-Clarity™ quality level = MQ200

8.07270.0100 8.07270.2500
EM8.07270.2500CS 1281 USD InStock
EM8.07270.2500 EM8.07270.0100
Phosphorus, red stabilized for synthesis, Sigma-Aldrich®
Phosphorus


Formula: P
MW: 30.97 g/mol
Density: ∼2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133771
CAS Number: 7723-14-0
EINECS: 231-768-7
UN: 1338
ADR: 4.1,III
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 590 °C
Molar Mass 30.97 g/mol
Bulk Density 1100 kg/m3
Vapor Pressure  - 0.1 hPa (20 °C)
Density 2.34 g/cm3 (20 °C)
pH 9.5 (100 g/l, H2O, 20 °C)
Ignition Point 260 °C
Hazard Class 4.1
Hazard Class Description Flammable Solid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty