Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isopropylamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: 2-Aminopropane

M-Clarity™ quality level = MQ200

8.07476.0100 8.07476.1000 8.07476.0100 8.07476.2500 8.07476.2500 8.07476.1000
EM8.07476.0100CS 808 USD InStock
CA8.07476.0100 CA8.07476.1000 EM8.07476.0100 EM8.07476.2500 EM8.07476.1000 CA8.07476.2500
Isopropylamine for synthesis, Sigma-Aldrich®
Isopropylamine


Formula: C₃H₉N
MW: 59.11 g/mol
Boiling Pt: 32.4 °C (1013 hPa)
Melting Pt: –101 °C
Density: 0.69 g/cm³ (20 °C)
Flash Pt: –27 °C
Storage Temperature: Ambient
MDL Number: MFCD00008082
CAS Number: 75-31-0
EINECS: 200-860-9
UN: 1221
ADR: 3,I
Merck Index: 13,05229
Order Now
Specification Test Results
Solubility 1000 g/l (20 °C)
Melting Point -101 °C
Molar Mass 59.11 g/mol
Boiling Point 31 - 33 °C (1013 hPa)
Vapor Pressure 632 hPa (20 °C)
Flash Point -37 °C
Refractive Index 1.3746 (20 °C, 589 nm)
Explosion Limit 2 - 10.4 %(V)
Density 0.69 g/cm3 (20 °C)
pH 13 (700 g/l, H2O, 20 °C)
Ignition Point 330 °C
Hazard Class 3
Hazard Class Description Flammable liquid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty