Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Pentanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: 1-Pentyl alcoholn-Amyl alcoholAmyl alcohol1-Hydroxy pentaneButyl carbinol

M-Clarity™ quality level = MQ200

8.07500.1000 8.07500.2500 8.07500.0100 8.07500.2500 8.07500.1000 8.07500.0100
EM8.07500.0100CS 650 USD InStock
EM8.07500.0100 EM8.07500.1000 CA8.07500.2500 CA8.07500.0100 CA8.07500.1000 EM8.07500.2500
1-Pentanol for synthesis, Sigma-Aldrich®
1-Pentanol


Formula: H₃C(CH₂)₃CH₂OH
MW: 88.15 g/mol
MDL Number: MFCD00002977
CAS Number: 71-41-0
UN: 1105
ADR: 3,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/15/15/731515.jpg
Order Now
Specification Test Results
Solubility 22 g/l (20 °C)
Melting Point -79 °C
Molar Mass 88.15 g/mol
Boiling Point 138 °C (1013 hPa)
Vapor Pressure 2.6 hPa (20 °C)
Flash Point 49 °C
Refractive Index 1.41 (20 °C, 589 nm)
Explosion Limit 1.6 %(V)
Density 0.815 g/cm3 (20 °C)
pH (22 g/l, H2O, 20 °C) neutral
Ignition Point 300 °C DIN 51794
Evaporation Number 2.7

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty