Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Octadecanol, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: Stearyl alcoholHeptadecyl carbinol1-Hydroxyoctadecane1-Octadecyl alcohol

M-Clarity™ quality level = MQ200

8.07680.1000 8.07680.0100 8.07680.1000 8.07680.0100
EM8.07680.1000CS 474 USD InStock
EM8.07680.1000 EM8.07680.0100 CA8.07680.1000 CA8.07680.0100
1-Octadecanol, Sigma-Aldrich®
1-Octadecanol
Stearyl alcohol for synthesis


Formula: H₃C(CH₂)₁₆CH₂OH
MW: 270.5 g/mol
MDL Number: MFCD00002823
CAS Number: 112-92-5
Order Now
Specification Test Results
Hill Formula C₁₈H₃₈O
Molar Mass 270.49 g/mol
Boiling point 330 - 360 °C (1013 hPa)
Density 0.805 - 0.815 g/cm³ (60 °C)
Flash point 195 °C
Ignition temperature 230 °C DIN 51794
Melting Point 55 - 60 °C
Vapor pressure  - 1 hPa (20 °C)
Bulk density 300 kg/m³
Solubility (20 °C) practically insoluble
LD 50 oral LD50 Rat 2000 mg/kg
Storage class 10 - 13 Other liquids and solids
WGK NWG not water endangering

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty