Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triisopropanolamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: 1,1',1"-Nitrilotri-2-propanol1,1',1''-Nitrilotripropan-2-olTris(2-hydroxypropyl)amine

M-Clarity™ quality level = MQ200

8.08236.1000 8.08236.0100 8.08236.1000 8.08236.0100
EM8.08236.1000CS 456 USD InStock
EM8.08236.1000 CA8.08236.0100 CA8.08236.1000 EM8.08236.0100
Triisopropanolamine for synthesis, Sigma-Aldrich®
Triisopropanolamine


Formula: C₉H₂₁NO₃
MDL Number: MFCD00004533
CAS Number: 122-20-3
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point 50 °C
Molar Mass 191.27 g/mol
Boiling Point 305 °C (1013 hPa)
Vapor Pressure 1 hPa (100 °C)
Flash Point 160 °C
Explosion Limit 0.8 - 5.8 %(V)
Density 1.01 g/cm3 (50 °C)
pH 10.8 (100 g/l, H2O, 20 °C)
Ignition Point 275 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty