Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triphenylphosphine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

M-Clarity™ quality level = MQ200

8.08270.0250 8.08270.1000 8.08270.0005 8.08270.0250 8.08270.1000 8.08270.0005
EM8.08270.1000CS 2194 USD InStock
EM8.08270.1000 EM8.08270.0005 CA8.08270.1000 CA8.08270.0250 EM8.08270.0250 CA8.08270.0005
Triphenylphosphine for synthesis, Sigma-Aldrich®
Triphenylphosphine


Formula: C₁₈H₁₅P
MW: 262.29 g/mol
Boiling Pt: 377 °C (1013 hPa)
Melting Pt: 79 °C
Density: 1.194 g/cm³ (20 °C)
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00003043
CAS Number: 603-35-0
EINECS: 210-036-0
Merck Index: 12,09873
Order Now
Specification Test Results
Solubility  - 0.0001 g/l (25 °C)
Melting Point 78.5 - 81.5 °C
Molar Mass 262.29 g/mol
Bulk Density 500 - 600 kg/m3
Boiling Point 195 - 205 °C (7 hPa)
Vapor Pressure 0.01 hPa (88 °C)
Flash Point 182 °C
Density 1.194 g/cm3 (20 °C)
Ignition Point 425 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty