Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrabutyltin for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: TetrabutylstannaneTetra-n-butyltin

M-Clarity™ quality level = MQ200

8.08278.0025 8.08278.0100 8.08278.0100 8.08278.0025
EM8.08278.0100CS 593 USD InStock
EM8.08278.0100 EM8.08278.0025 CA8.08278.0100 CA8.08278.0025
Tetrabutyltin for synthesis, Sigma-Aldrich®
Tetrabutyltin


Formula: C₁₆H₃₆Sn
MW: 347.17 g/mol
Boiling Pt: 145 °C (1013 hPa)
Melting Pt: –97 °C
Density: 1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009422
CAS Number: 1461-25-2
EINECS: 215-960-8
UN: 2788
ADR: 6.1,III
Order Now
Specification Test Results
Solubility 0.008 g/l (20 °C)
Melting Point -97 °C
Molar Mass 347.15 g/mol
Boiling Point 140 °C (10 hPa)
Vapor Pressure 0.0014 hPa (25 °C)
Flash Point 100 °C
Refractive Index 1.4742 (20 °C, 589 nm)
Density 1.05 g/cm3 (20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty