Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl toluene-4-sulfonate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: Ethyl p-toluenesulfonatep-Toluenesulphonic acid ethyl esterEthyl 4-toluenesulfonateEthyl 4-methylbenzenesulphonateToluene-4-sulphonic acid ethyl esterEthyl 4-methylbenzenesulfonateEthyl p-toluenesulphonatep-Toluenesulfonic acid ethyl esterToluene-4-sulfonic acid ethyl esterEthyl 4-toluenesulphonate

M-Clarity™ quality level = MQ200

8.08332.0500 8.08332.0100 8.08332.5000 8.08332.0100 8.08332.5000 8.08332.0500
EM8.08332.5000EA 454 USD InStock
CA8.08332.0500 EM8.08332.5000 CA8.08332.0100 EM8.08332.0500 CA8.08332.5000 EM8.08332.0100
Ethyl toluene-4-sulfonate for synthesis, Sigma-Aldrich®
Ethyl toluene-4-sulphonate


Formula: C₉H₁₂O₃S
MW: 200.26 g/mol
MDL Number: MFCD00009100
CAS Number: 80-40-0
ADR: 6.1,III
Order Now
Specification Test Results
Solubility 1.24 g/l (25 °C)
Melting Point 30 - 32 °C
Molar Mass 200.25 g/mol
Boiling Point 221 °C (1013 hPa)
Vapor Pressure 0.6 hPa (14 °C)
Flash Point 158 °C
Density 1.174 g/cm3 (20 °C)
Ignition Point 500 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty