Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tris(2-hydroxyethyl)-1,3,5-triazinetrione for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: 1,3,5-Tris(2-hydroxyethyl)isocyanuric acid1,3,5-Tris(2-hydroxyethyl)-1,3,5-triazinane-2,4,6-trione1,3,5-Tris(2-hydroxyethyl)cyanuric acid1,3,5-Tris(2-hydroxyethyl)isocyanurate

M-Clarity™ quality level = MQ200

8.08608.0250 8.08608.0250
EM8.08608.0250CS 7381 USD InStock
CA8.08608.0250 EM8.08608.0250
Tris(2-hydroxyethyl)-1,3,5-triazinetrione for synthesis, Sigma-Aldrich®
Tris(2-hydroxyethyl)-1,3,5-triazinetrione


Formula: C₉H₁₅N₃O₆
MW: 261.23 g/mol
MDL Number: MFCD00003549
CAS Number: 839-90-7
Order Now
Specification Test Results
Solubility 1150 g/l (20 °C)
Melting Point 133 - 136 °C
Molar Mass 261.23 g/mol
Bulk Density 580 kg/m3
Flash Point 270 °C
pH 7 (100 g/l, H2O, 20 °C)
Ignition Point 430 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty