Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,2,2-Trichloroethanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: 2,2,2-Trichloroethan-1-ol

M-Clarity™ quality level = MQ200

8.08610.0100
EM8.08610.0100CS 2435 USD InStock
EM8.08610.0100
2,2,2-Trichloroethanol for synthesis, Sigma-Aldrich®
2,2,2-Trichloroethanol


Formula: C₂H₃Cl₃O
MW: 149.4 g/mol
MDL Number: MFCD00004677
CAS Number: 115-20-8
ADR: 6.1,III
Order Now
Specification Test Results
Solubility 83 g/l (20 °C)
Melting Point 17 - 18 °C
Molar Mass 149.4 g/mol
Boiling Point 151 - 153 °C (1013 hPa)
Vapor Pressure 1.3 hPa (20 °C)
Flash Point 110 °C
Refractive Index 1.4885 (20 °C, 589 nm)
Density 1.56 g/cm3 (20 °C)
pH 5 - 6 (H2O, 20 °C) (saturated solution)
Ignition Point 151 °C
Synonyms ß,ß,ß-Trichloroethyl alcohol
Hill Formula C2H3Cl3O
Chemical Formula CCl3CH2OH

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty