Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

p-Xylene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: 1,4-Dimethylbenzene

M-Clarity™ quality level = MQ200

8.08691.2500 8.08691.1000 8.08691.2500 8.08691.1000
EM8.08691.1000CS 469 USD InStock
EM8.08691.1000 EM8.08691.2500 CA8.08691.1000 CA8.08691.2500
p-Xylene for synthesis, Sigma-Aldrich®
p-Xylene


Formula: C₆H₄(CH₃)₂
MW: 106.17 g/mol
MDL Number: MFCD00008556
CAS Number: 106-42-3
UN: 1307
ADR: 3,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/87/75/768775.jpg
Order Now
Specification Test Results
Solubility 0.2 g/l (25 °C)
Melting Point 13.3 °C
Molar Mass 106.17 g/mol
Boiling Point 138.4 °C (1013 hPa)
Vapor Pressure 8.7 hPa (20 °C)
Flash Point 24 °C
Explosion Limit 1.1 - 7 %(V)
Density 0.86 g/cm3 (20 °C)
Ignition Point 540 °C
Evaporation Number 13.5

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty