Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc(II) acetylacetonate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: Zinc (II) 2,4-pentanedionateZinc (II) bis(2,4-pentanedionato-O,O')Bis(2,4-pentanedionato-O,O')zinc (II)Zinc acetylacetonate

M-Clarity™ quality level = MQ200

8.08803.0100 8.08803.0100
EM8.08803.0100CS 1580 USD InStock
CA8.08803.0100 EM8.08803.0100
Zinc(II) acetylacetonate for synthesis, Sigma-Aldrich®
Zinc (II) acetylacetonate


Formula: C₁₀H₁₄O₄Zn
MW: 263.61 g/mol
MDL Number: MFCD00000035
CAS Number: 14024-63-6
Order Now
Specification Test Results
Solubility 6.9 g/l (20 °C)
Melting Point 124 - 126 °C
Molar Mass 263.61 g/mol
Bulk Density 500 kg/m3
Boiling Point 129 - 131 °C (13 hPa)
Vapor Pressure 1 hPa (20 °C)
pH 10 (4 g/l, H2O, 20 °C)
Ignition Point 580 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty