Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dibenzo-24-crown-8-(dibenzo-1,4,7,10,13,16,19,22-octaoxacyclotetracosane) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: Dibenzo-24-crown-8

M-Clarity™ quality level = MQ200

8.13755.0001 8.13755.0005 8.13755.0005 8.13755.0001
EM8.13755.0005CS 431827 USD InStock
CA8.13755.0005 EM8.13755.0005 EM8.13755.0001 CA8.13755.0001
Dibenzo-24-crown-8-(dibenzo-1,4,7,10,13,16,19,22-octaoxacyclotetracosane) for synthesis, Sigma-Aldrich®
Dibenzo-24-crown-8 (Dibenzo-1,4,7,10,13,16,19,22-octaoxacyclotetracosane)


Formula: C₂₄H₃₂O₈
MW: 448.51 g/mol
MDL Number: MFCD00005101
CAS Number: 14174-09-5
Order Now
Specification Test Results
Melting Point 100 - 103 °C
Molar Mass 448.52 g/mol

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty