Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N'-Dibenzylethylenediamine diacetate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: Benzathine diacetateN1,N2-Dibenzylethane-1,2-diamine diacetateN,N-Dibenzylethylenediamine diacetate

M-Clarity™ quality level = MQ200

8.14050.0100 8.14050.0100
EM8.14050.0100CS 757 USD InStock
EM8.14050.0100 CA8.14050.0100
N,N'-Dibenzylethylenediamine diacetate for synthesis, Sigma-Aldrich®
N,N'-Dibenzylethylenediamine diacetate


Formula: C₁₆H₂₀N₂
MW: 360.45 g/mol
MDL Number: MFCD00040588
CAS Number: 122-75-8
Order Now
Specification Test Results
Solubility 150 g/l (20 °C)
Melting Point 118 °C
Molar Mass 360.45 g/mol
Bulk Density 280 kg/m3
pH 5 - 6 (50 g/l, H2O, 20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty