Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-(+)-2-Amino-3-methyl-1-butanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: L-ValinolL(+)-ValinolL(+)-2-Amino-3-methyl-1-butanol(S)-2-Amino-3-methylbutan-1-ol(S)-2-Amino-3-methyl-butan-1-ol

M-Clarity™ quality level = MQ200

8.14082.0001 8.14082.0001
EM8.14082.0001CS 2555 USD InStock
EM8.14082.0001 CA8.14082.0001
(S)-(+)-2-Amino-3-methyl-1-butanol for synthesis, Sigma-Aldrich®
(S)-(+)-2-Amino-3-methyl-1-butanol


Formula: C₅H₁₃NO
MW: 103.16 g/mol
MDL Number: MFCD00064296
CAS Number: 2026-48-4
UN: 2811
ADR: 6.1,II
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 31 - 32 °C
Molar Mass 103.16 g/mol
Boiling Point 91 - 92 °C (13 hPa)
Flash Point 78 °C
Refractive Index 1.4548 (20 °C, 589 nm)
Density 0.93 g/cm3 (20 °C)
Synonyms L-Valinol
Hill Formula C5H13NO
Chemical Formula CH3(CH3)CHCH(NH2)CH2OH

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty