Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tributyltin hydride, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: Tri-n-butyltin hydride

M-Clarity™ quality level = MQ200

8.14109.0005 8.14109.0025 8.14109.0100 8.14109.0025 8.14109.0100 8.14109.0005
EM8.14109.0100CS 5274 USD InStock
CA8.14109.0005 EM8.14109.0100 EM8.14109.0025 EM8.14109.0005 CA8.14109.0100 CA8.14109.0025
Tributyltin hydride, Sigma-Aldrich®
Tributyltin hydride
Tributyltin hydride for synthesis


Formula: C₁₂H₂₈Sn
MW: 291.06 g/mol
MDL Number: MFCD00009416
CAS Number: 688-73-3
UN: 2788
ADR: 6.1,II
Order Now
Specification Test Results
Hill Formula C₁₂H₂₈Sn
Molar Mass 291.06 g/mol
Boiling point 112 - 114 °C (11 hPa)
Density 1.10 g/cm³ (20 °C)
Flash point 109 °C
Ignition temperature 250 °C
Vapor pressure 5 hPa (20 °C)
Refractive Index 1.4726 (20 °C, 589 nm)
Storage class 6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK WGK 3 highly water endangering
R 21-25-36/38-48/23/25-50/53 S Phrase
S 35-36/37/39-45-60-61

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty