Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(Dimethylamino)benzoic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

M-Clarity™ quality level = MQ200

8.14112.0025 8.14112.0025
EM8.14112.0025CS 26539 USD InStock
CA8.14112.0025 EM8.14112.0025
3-(Dimethylamino)benzoic acid for synthesis, Sigma-Aldrich®
3-(Dimethylamino)benzoic acid


Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD00002497
CAS Number: 99-64-9
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 149 - 152 °C
Molar Mass 165.19 g/mol
Hill Formula C9H11NO2
Chemical Formula 3-((CH3)2N)C6H4COOH

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty