Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Polyethylene glycol dimethylether, M.W. 2,000 for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

M-Clarity™ quality level = MQ200

8.14172.0500 8.14172.0500
EM8.14172.0500CS 1665 USD InStock
EM8.14172.0500 CA8.14172.0500
Polyethylene glycol dimethylether, M.W. 2,000 for synthesis, Sigma-Aldrich®
Polyethylene glycol dimethylether 150


Formula: H₃C(OCH₂CH₂)nOCH₃
MDL Number: MFCD00147972
CAS Number: 24991-55-7
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point 49 - 53 °C
Molar Mass 2000 g/mol
Boiling Point 250 °C (1013 hPa)
Vapor Pressure  - 0.01 hPa (20 °C)
Flash Point 254 °C
Refractive Index 1.455 (60 °C, 589 nm)
Density 1.08 g/cm3 (60 °C)
pH (100 g/l, H2O, 20 °C) neutral
Ignition Point 385 °C DIN 51794
Viscosity kinematic 30 mm2/s (100 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty