Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hydroxide, powder for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: Potassium hydrateCaustic potash

M-Clarity™ quality level = MQ200

8.14353.5000 8.14353.1000 8.14353.1000 8.14353.9050 8.14353.9050 8.14353.0100 8.14353.0100 8.14353.5000
EM8.14353.5000CS 1321 USD InStock
EM8.14353.5000 CA8.14353.0100 CA8.14353.1000 CA8.14353.5000 EM8.14353.9050 EM8.14353.0100 EM8.14353.1000 CA8.14353.9050
Potassium hydroxide, powder for synthesis, Sigma-Aldrich®
Potassium hydroxide


Formula: KOH
MW: 56.11 g/mol
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1813
ADR: 8,II
Order Now
Specification Test Results
Solubility 1130 g/l (20 °C)
Melting Point 360 °C
Molar Mass 56.11 g/mol
Boiling Point 1320 °C
Vapor Pressure (20 °C)
Density 2.04 g/cm3 (20 °C)
pH 14 (56 g/l, H2O, 20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty