Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(1S)-(-)-3-Oxocamphorsulfonylimine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: (1S,7S)-10,10-Dimethyl-3-thia-4-azatricyclo[5.2.1.01,5]dec-4-en-6-one 3,3-dioxide3H-3a,6-Methano-2,1-benzisothiazol-7(4H)-one, 5,6-dihydro-8,8-dimethyl-, 2,2-dioxide, (3aS,6S)-

M-Clarity™ quality level = MQ200

8.14388.0001 8.14388.0005 8.14388.0005 8.14388.0001
EM8.14388.0005CS 17891 USD InStock
EM8.14388.0005 CA8.14388.0001 CA8.14388.0005 EM8.14388.0001
(1S)-(-)-3-Oxocamphorsulfonylimine for synthesis, Sigma-Aldrich®
(1S)-(-)-3-Oxocamphorsulfonylimine


Formula: C₁₀H₁₃NO₃S
MW: 227.28 g/mol
MDL Number: MFCD00066268
CAS Number: 119106-38-6
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 195 - 198 °C
Molar Mass 227.28 g/mol
Hill Formula C10H13NO3S

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty