Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloromercuriferrocene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: Iron(2+) chloro(3,5-cyclopentadien-2-id-1-yl)mercury 2,4-cyclopentadienide (1:1:1)FERROCENEMERCURY(II) CHLORIDE

M-Clarity™ quality level = MQ200

8.14400.0001 8.14400.0001
EM8.14400.0001CS 164844 USD InStock
EM8.14400.0001 CA8.14400.0001
Chloromercuriferrocene for synthesis, Sigma-Aldrich®
Chloromercuriferrocene


Formula: C₁₀H₉ClFeHg
MW: 421.07 g/mol
MDL Number: MFCD00013751
CAS Number: 1273-75-2
UN: 2025
ADR: 6.1,II
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Molar Mass 421.07 g/mol

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty