Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

O-Phenylenediamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: 1,2-PhenylendiamineOPD1,2-Diaminobenzene1,2-Phenylenediamine

M-Clarity™ quality level = MQ200

8.14538.0250 8.14538.0005 8.14538.0005 8.14538.1000 8.14538.1000 8.14538.0250
EM8.14538.1000CS 836 USD InStock
EM8.14538.1000 CA8.14538.0005 EM8.14538.0250 CA8.14538.0250 EM8.14538.0005 CA8.14538.1000
O-Phenylenediamine for synthesis, Sigma-Aldrich®
o-Phenylenediamine


Formula: C₆H₈N₂
MW: 108.14 g/mol
MDL Number: MFCD00007721
CAS Number: 95-54-5
UN: 1673
ADR: 6.1,III
Order Now
Specification Test Results
Solubility 54 g/l (20 °C)
Melting Point 99 - 102 °C
Molar Mass 108.14 g/mol
Bulk Density 530 kg/m3
Boiling Point 257 °C (1013 hPa)
Vapor Pressure 0.013 hPa (20 °C)
Flash Point 110 °C
Explosion Limit 1.5 %(V)
Density 1.14 g/cm3 (20 °C)
pH 7 - 8 (50 g/l, H2O, 20 °C)
Ignition Point 540 °C
Hazard Class 6.1
Hazard Class Description Toxic materials

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty