Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Propylcyclohexanone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

M-Clarity™ quality level = MQ200

8.14584.0250 8.14584.0250 8.14584.0005 8.14584.0005
EM8.14584.0250CS 5929 USD InStock
EM8.14584.0250 CA8.14584.0005 EM8.14584.0005 CA8.14584.0250
4-Propylcyclohexanone for synthesis, Sigma-Aldrich®
4-Propylcyclohexanone


Formula: C₉H₁₆O
MW: 140.23 g/mol
MDL Number: MFCD00058690
CAS Number: 40649-36-3
Order Now
Specification Test Results
Solubility 1.96 g/l (20 °C)
Molar Mass 140.22 g/mol
Boiling Point 219 °C (1013 hPa)
Vapor Pressure 0.23 hPa (20 °C)
Flash Point 90 °C
Density 0.91 g/cm3 (20 °C)
pH 6.0 (2 g/l, H2O, 20 °C)
Ignition Point 320 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty