Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(II) bromide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: Cuprous bromideCupric bromide

M-Clarity™ quality level = MQ200

8.14658.0250 8.14658.0250 8.14658.0050 8.14658.0050
EM8.14658.0050CS 1761 USD InStock
EM8.14658.0050 EM8.14658.0250 CA8.14658.0050 CA8.14658.0250
Copper(II) bromide for synthesis, Sigma-Aldrich®
Copper (II) bromide


Formula: Br₂Cu
MW: 223.35 g/mol
MDL Number: MFCD00010970
CAS Number: 7789-45-9
UN: 1759
ADR: 8,III
Order Now
Specification Test Results
Solubility 1200 g/l (25 °C)
Melting Point 498 °C
Molar Mass 223.35 g/mol
Bulk Density 2000 kg/m3
Boiling Point 900 °C (1013 hPa)
Density 4.71 g/cm3 (20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty