Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methyl-2-butene mixture of isomers for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: TrimethylethyleneIsoamyleneIsopentene

M-Clarity™ quality level = MQ200

8.14874.0250 8.14874.9025 8.14874.0025 8.14874.1000 8.14874.0025 8.14874.9025 8.14874.0250 8.14874.1000
EM8.14874.0025CS 5595 USD InStock
CA8.14874.0250 EM8.14874.0025 CA8.14874.1000 EM8.14874.1000 EM8.14874.9025 EM8.14874.0250 CA8.14874.9025 CA8.14874.0025
2-Methyl-2-butene mixture of isomers for synthesis, Sigma-Aldrich®
2-Methyl-2-butene


Formula: C₅H₁₀
MW: 70.13 g/mol
MDL Number: MFCD00009276
CAS Number: 513-35-9
UN: 3295
ADR: 3,II
Order Now
Specification Test Results
Solubility 0.19 g/l (25 °C)
Melting Point -134 °C
Molar Mass 70.13 g/mol
Vapor Pressure 513 hPa (20 °C)
Flash Point -45 °C
Explosion Limit 1 - 9 %(V)
Density 0.66 g/cm3 (20 °C)
Ignition Point 290 °C
Viscosity Kinematic 0.29 mm2/s (20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty