Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-one for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: 1,3-Dimethyltetrahydropyrimidin-2(1H)-oneDimethyl propylene urea1,3-Dimethyltetrahydro-2(1H)-pyrimidinone1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinoneN,N'-Dimethyl propylene ureaDMPU

M-Clarity™ quality level = MQ200

8.18214.9025 8.18214.0100 8.18214.9025 8.18214.0500 8.18214.0500 8.18214.0100
EM8.18214.0500CS 1372 USD InStock
EM8.18214.0500 CA8.18214.0100 CA8.18214.9025 CA8.18214.0500 EM8.18214.0100 EM8.18214.9025
Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-one for synthesis, Sigma-Aldrich®
Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-one


Formula: C₆H₁₂N₂O
MW: 128.17 g/mol
MDL Number: MFCD00006550
CAS Number: 7226-23-5
Order Now
Specification Test Results
Solubility (20°C) Soluble
Melting Point -24 °C
Boiling Point 247 °C (1013 hPa)
Vapor Pressure 0.5 hPa (57 °C)
Flash Point 121 °C
Explosion Limit 1.6 - 9.3 %(V)
Density 1.064 g/cm3 (20 °C)
pH 11 (100 g/l, H2O, 20 °C)
Ignition Point 255°C
Synonyms Dimethylpropyleneurea, DMPU
Hill Formula C6H12N2O

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty