Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Semicarbazide hydrochloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: Hydrazinecarboxamide hydrochloride

M-Clarity™ quality level = MQ200

8.18369.0100 8.18369.0100 8.18369.0500 8.18369.0500
EM8.18369.0100CS 581 USD InStock
CA8.18369.0500 EM8.18369.0100 EM8.18369.0500 CA8.18369.0100
Semicarbazide hydrochloride for synthesis, Sigma-Aldrich®
Semicarbazide hydrochloride


Formula: CH₆ClN₃O
MW: 111.53 g/mol
MDL Number: MFCD00013009
CAS Number: 563-41-7
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point 175 - 177 °C (decomposition)
Molar Mass 111.53 g/mol
Bulk Density 740 kg/m3
pH 1.5 (100 g/l, H2O, 20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty